2,4,6-Trimethylphenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
T79006-100G T79006-25G
SIALT79006-100GEA 240 EUR
SIALT79006-100G SIALT79006-25G
2,4,6-Trimethylphenol, Sigma-Aldrich®
2,4,6-Trimethylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 56 °C (0.1 mmHg)
Melting Pt: 74
Density: 1.046 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002235
CAS Number: 527-60-6
EINECS: 208-419-2
UN: 2430
ADR: 8 III