2,3,5-Trimethylhydroquinone, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2,3,5-Trimethylbenzene-1,4-diol, Trimethylhydroquinone

M-Clarity™ quality level = MQ100

T76503-100G T76503-500G
SIALT76503-100GEA 55.9 EUR
SIALT76503-100G SIALT76503-500G
2,3,5-Trimethylhydroquinone, Sigma-Aldrich®
2,3,5-Trimethylhydroquinone
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00002346
CAS Number: 700-13-0
EINECS: 211-838-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR