1-Iodo-2,3,5,6-tetramethylbenzene

Supplier: Thermo Scientific

Synonyms: 2,3,5,6-Tetramethyliodobenzene, 3-Iodo-1,2,4,5-tetramethylbenzene

CD07686.1 CD07686.10 CD07686.50
MAYBCD07686.1EA 0 EUR
MAYBCD07686.1 MAYBCD07686.10 MAYBCD07686.50
1-Iodo-2,3,5,6-tetramethylbenzene
1-Iodo-2,3,5,6-tetramethylbenzene
Formula: C₁₀H₁₃I
MW: 260.12 g/mol
MDL Number: MFCD00013705
CAS Number: 2100-25-6
EINECS: 218-266-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR