3-Bromobiphenyl (PBB No. 2), Sigma-Aldrich®
Supplier: Merck
Synonyms:
PBB No. 2
M-Clarity™ quality level = MQ100
255386-1G
255386-5G
SIAL255386-1GEA
26.1
EUR
SIAL255386-1G
SIAL255386-5G
3-Bromobiphenyl (PBB No. 2), Sigma-Aldrich®
3-Bromobiphenyl (PBB No. 2)
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 299 °C (1013 hPa) Melting Pt: 9 °C Density: 1.41 g/cm³ (20 °C) |
MDL Number:
MFCD00000082 CAS Number: 2113-57-7 EINECS: 218-304-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...