4-Bromobiphenyl (PBB No. 3), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl
M-Clarity™ quality level = MQ100
281999-10G 281999-50G
SIAL281999-10GEA 29.3 EUR
SIAL281999-10G SIAL281999-50G
4-Bromobiphenyl (PBB No. 3), Sigma-Aldrich®
4-Bromobiphenyl (PBB No. 3)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 90…91 °C
Density: 1.615 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000100
CAS Number: 92-66-0
EINECS: 202-176-6