3,3',4-Trichlorobiphenyl (PCB No. 35)

Supplier: EHRENSTORFER GMBH

EHERC20003500EA 111 EUR
EHERC20003500
3,3',4-Trichlorobiphenyl (PCB No. 35)
3,3',4-Trichlorobiphenyl (PCB No. 35)
Formula: C₁₂H₇Cl₃
MW: 257,55 g/mol
UN: 3432
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR