Benzyl benzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzoic acid benzyl ester
B6630-1L B6630-250ML B6630-500ML
SIALB6630-1LEA 117 EUR
SIALB6630-1L SIALB6630-250ML SIALB6630-500ML
Benzyl benzoate, Sigma-Aldrich®
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 17…20 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129