Benzyl benzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzoic acid benzyl ester
M-Clarity™ quality level = MQ200
B9550-1L B9550-250ML B9550-500ML
SIALB9550-1LEA 182 EUR
SIALB9550-1L SIALB9550-250ML SIALB9550-500ML
Benzyl benzoate, Sigma-Aldrich®
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 17…20 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129