Benzyl butyl phthalate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: BBP

M-Clarity™ quality level = MQ100

308501-1L 308501-250ML 308501-5ML
SIAL308501-1LEA 217 EUR
SIAL308501-1L SIAL308501-250ML SIAL308501-5ML
Benzyl butyl phthalate, Sigma-Aldrich®
Benzyl butyl phthalate
Formula: C₁₉H₂₀O₄
Boiling Pt: 370 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009440
CAS Number: 85-68-7
EINECS: 201-622-7
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR