4-Phenylaniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl
M-Clarity™ quality level = MQ200
A2898-1G A2898-5G
SIALA2898-1GEA 43.3 EUR
SIALA2898-1G SIALA2898-5G
4-Phenylaniline, Sigma-Aldrich®
4-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 55 °C
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00007879
CAS Number: 92-67-1
EINECS: 202-177-1
Merck Index: 12,01276