4,4'-Biphenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: [1,1'-Biphenyl]-4,4'-diol, 4,4'-Dihydroxybiphenyl, Biphenyl-4,4'-diol
M-Clarity™ quality level = MQ100
168734-100G 168734-25G
SIAL168734-100GEA 335 EUR
SIAL168734-100G SIAL168734-25G
4,4'-Biphenol, Sigma-Aldrich®
4,4'-Biphenol
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 355 °C (1013 hPa)
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00002348
CAS Number: 92-88-6
EINECS: 202-200-5