Biphenyl (PCB No. 0), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB
M-Clarity™ quality level = MQ300
W312908-10KG W312908-1KG W312908-SAMPLE
SIALW312908-10KGEA 177 EUR
SIALW312908-10KG SIALW312908-1KG SIALW312908-SAMPLE
Biphenyl (PCB No. 0), Sigma-Aldrich®
Biphenyl (PCB No. 0)
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 255
Melting Pt: 68…70 °C
Density: 1.175 g/cm³ (20 °C)
Flash Pt: 113
Storage Temperature: Ambient
MDL Number: MFCD00003054
CAS Number: 92-52-4
EINECS: 202-163-5
UN: 3077
ADR: 9 III
Merck Index: 13,03346