Coumarin, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone
M-Clarity™ quality level = MQ200
C4261-100G C4261-500G C4261-50G
SIALC4261-100GEA 55.3 EUR
SIALC4261-100G SIALC4261-500G SIALC4261-50G
Coumarin, Sigma-Aldrich®
Coumarin
Formula: C₉H₆O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 67…69 °C
Density: 0.935 g/cm³ (20 °C)
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588