7-Methoxycoumarin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
7-Methoxy-2-benzopyrone, 7-Methoxy-2H-chromen-2-one
M-Clarity™ quality level = MQ300
W515809-25G
W515809-SAMPLE
SIALW515809-25GEA
144
EUR
SIALW515809-25G
SIALW515809-SAMPLE
7-Methoxycoumarin, Sigma-Aldrich®
7-Methoxycoumarin
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 71 °C Density: 1.36 g/cm³ (20 °C) |
MDL Number:
MFCD00006876 CAS Number: 531-59-9 EINECS: 208-513-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...