7-Hydroxycoumarin, Sigma-Aldrich®

Supplier: Merck

Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone

M-Clarity™ quality level = MQ100

H24003-100G H24003-10G H24003-25G
SIALH24003-100GEA 152 EUR
SIALH24003-100G SIALH24003-10G SIALH24003-25G
7-Hydroxycoumarin, Sigma-Aldrich®
7-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 382 °C (1013 hPa)
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00006878
CAS Number: 93-35-6
EINECS: 202-240-3
Merck Index: 12,09981

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR