7-Amino-4-methylcoumarin 98%
Supplier: Thermo Scientific
Warning
Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
A15017.03 A15017.06 A15017.MD
A15017.03EA 125 EUR
A15017.03 A15017.06 A15017.MD
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin
Purity: 98% CAS No.: 26093-31-2 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 225-228° Beilstein Reference: 142231
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 225…228 °C
Density: 1.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8