7-Amino-4-methylcoumarin
Supplier: Molekula
Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
22061681-1G 22061681-5G
MOLE22061681-1GEA 52.8 EUR
MOLE22061681-1G MOLE22061681-5G
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
Density: 1.377 g/cm³ (20 °C)
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8