7-Amino-4-methylcoumarin, Sigma-Aldrich®
Supplier: Merck
Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
257370-100MG 257370-500MG
SIAL257370-100MGEA 72.9 EUR
SIAL257370-100MG SIAL257370-500MG
7-Amino-4-methylcoumarin, Sigma-Aldrich®
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
Density: 1.377 g/cm³ (20 °C)
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8