Benzo[ghi]perylene 98+%
Supplier: Thermo Fisher Scientific
Warning
105551000 105550050 105550250
ACRO105551000EA 395 EUR
ACRO105551000 ACRO105550050 ACRO105550250
Benzo[ghi]perylene 98+%
Benzo[ghi]perylene
Formula: C₂₂H₁₂
MW: 276.34 g/mol
Boiling Pt: >500 °C (760 mmHg)
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004135
CAS Number: 191-24-2
EINECS: 205-883-8
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 276 to 280 °C
GC-MS ≥98.0 %