(1R)-(+)-α-Pinene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (+)-α-Pinene, D(+)-α-Pinene, (1R)-(+)-2-Pinene, (+)-Pin-2(3)-ene
M-Clarity™ quality level = MQ100
268070-25G 268070-5G
SIAL268070-25GEA 176 EUR
SIAL268070-25G SIAL268070-5G
(1R)-(+)-α-Pinene, Sigma-Aldrich®
(1R)-(+)-α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.857 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001346
CAS Number: 7785-70-8
EINECS: 232-087-8
UN: 2368
ADR: 3,III
Merck Index: 12,07599