(1S)-(-)-α-Pinene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
M-Clarity™ quality level = MQ100
305715-25G 305715-5G
SIAL305715-25GEA 122 EUR
SIAL305715-25G SIAL305715-5G
(1S)-(-)-α-Pinene, Sigma-Aldrich®
(1S)-(-)-α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.8592 g/cm³ (20 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064145
CAS Number: 7785-26-4
EINECS: 232-077-3
UN: 2368
ADR: 3,III
Merck Index: 12,07599