3-Bromoquinoline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
124133-100G 124133-25G
SIAL124133-100GEA 278 EUR
SIAL124133-100G SIAL124133-25G
3-Bromoquinoline, Sigma-Aldrich®
3-Bromoquinoline
Formula: C₉H₆BrN
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006767
CAS Number: 5332-24-1
EINECS: 226-237-1