2-Chloroquinoline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C70401-1G C70401-25G C70401-5G
SIALC70401-1GEA 65 EUR
SIALC70401-1G SIALC70401-25G SIALC70401-5G
2-Chloroquinoline, Sigma-Aldrich®
2-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 266…267 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1.2464 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006741
CAS Number: 612-62-4
EINECS: 210-317-8