4-Chloroquinoline, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

C70509-1G C70509-5G
SIALC70509-1GEA 67.6 EUR
SIALC70509-1G SIALC70509-5G
4-Chloroquinoline, Sigma-Aldrich®
4-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 260…261 °C (1013 hPa)
Melting Pt: 28
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006773
CAS Number: 611-35-8
EINECS: 210-267-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR