5,7-Dichloro-8-quinolinol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Chloroxine, 5,7-Dichloro-8-hydroxyquinoline
M-Clarity™ quality level = MQ100
D64600-100G D64600-5G
SIALD64600-100GEA 88.7 EUR
SIALD64600-100G SIALD64600-5G
5,7-Dichloro-8-quinolinol, Sigma-Aldrich®
5,7-Dichloro-8-quinolinol
Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 183 °C
MDL Number: MFCD00006786
CAS Number: 773-76-2
EINECS: 212-258-3
Merck Index: 13,02193