5-Aminoquinoline, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Quinolin-5-amine, Quinolin-5-ylamine
M-Clarity™ quality level = MQ100
A79205-1G A79205-5G
SIALA79205-1GEA 79.4 EUR
SIALA79205-1G SIALA79205-5G
5-Aminoquinoline, Sigma-Aldrich®
5-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7
EINECS: 210-266-1