6-Aminoquinoline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Quinolin-6-ylamine
M-Clarity™ quality level = MQ200
275581-1G
275581-5G
SIAL275581-1GEA
50.6
EUR
SIAL275581-1G
SIAL275581-5G
6-Aminoquinoline, Sigma-Aldrich®
6-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 376 °C (1013 hPa) Melting Pt: 116…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006803 CAS Number: 580-15-4 EINECS: 209-453-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...