2-Quinolinol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Quinolin-2(1H)-one, Quinolin-2-ol, Carbostyril, 2-Quinolone, 2-Hydroxyquinoline
M-Clarity™ quality level = MQ100
270873-1G 270873-5G
SIAL270873-1GEA 51.5 EUR
SIAL270873-1G SIAL270873-5G
2-Quinolinol, Sigma-Aldrich®
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 199…200 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835