4-Quinolinol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Quinolin-4-ol, 4-Hydroxyquinoline
M-Clarity™ quality level = MQ100
H58005-10G H58005-1G H58005-5G
SIALH58005-10GEA 163 EUR
SIALH58005-10G SIALH58005-1G SIALH58005-5G
4-Quinolinol, Sigma-Aldrich®
4-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
EINECS: 210-268-2