Quinoline, Sigma-Aldrich®

Supplier: Merck
Danger


MQ200

241571-100G 241571-500G 241571-5G
SIAL241571-100GEA 63.5 EUR
SIAL241571-100G SIAL241571-500G SIAL241571-5G
Quinoline, Sigma-Aldrich®
Quinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 238
Melting Pt: –15 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 107 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1 III
Merck Index: 13,08160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e452fdc6c4f70d31bc2cfb5dade041787f8be427&vid=ec752804bedb2e9f8adfbea9ad287c2452f87a17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR