4-Methylquinoline, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Lepidine
MQ100
158283-100G 158283-25G
SIAL158283-100GEA 177 EUR
SIAL158283-100G SIAL158283-25G
4-Methylquinoline, Sigma-Aldrich®
4-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 261
Melting Pt: 9
Density: 1.0995 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0
EINECS: 207-734-2
Merck Index: 13,05460