6-Methylquinoline, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W274402-100G W274402-1KG W274402-SAMPLE
SIALW274402-100GEA 235 EUR
SIALW274402-100G SIALW274402-1KG SIALW274402-SAMPLE
6-Methylquinoline, Sigma-Aldrich®
6-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 258.6 °C (1013 hPa)
Melting Pt: –22.2 °C
Density: 1.0815 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006804
CAS Number: 91-62-3
EINECS: 202-084-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR