5-Methylquinoxaline 98
Supplier: Thermo Fisher Scientific
|
Warning
|
A19300.03
A19300.06
A19300.03EA
24.8
EUR
A19300.03
A19300.06
5-Methylquinoxaline 98
5-Methylquinoxaline
Purity: 98%
CAS No.: 13708-12-8
Molecular Formula: C9H8N2
Formula Weight: 144.18
Flash Point:
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 119…121 °C (15 mmHg) Melting Pt: 20…21 °C Density: 1.12 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012335 CAS Number: 13708-12-8 EINECS: 237-246-5 |