Vitamin B12 (cyanocobalamin), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
V6629-100MG V6629-1G V6629-250MG V6629-5G
SIALV6629-100MGEA 77.2 EUR
SIALV6629-100MG SIALV6629-1G SIALV6629-250MG SIALV6629-5G
Vitamin B12 (cyanocobalamin), Sigma-Aldrich®
Vitamin B12 (cyanocobalamin)
Formula: C₆₃H₈₈CoN₁₄O₁₄P
MW: 1355.38 g/mol
Melting Pt: 320
Storage Temperature: Refrigerator
MDL Number: MFCD00151092
CAS Number: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152