p-Benzoquinone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
M-Clarity™ quality level = MQ200
B10358-100G B10358-500G B10358-5G B10358-1KG
SIALB10358-100GEA 48.7 EUR
SIALB10358-100G SIALB10358-500G SIALB10358-5G SIALB10358-1KG
p-Benzoquinone, Sigma-Aldrich®
p-Benzoquinone
Formula: C₆H₄O₂
MW: 108.1 g/mol
Boiling Pt: ∼174 °C (1013 hPa)
Melting Pt: 112…114 °C
Density: 1.318 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=759b78f70866f47baad25776f322417b64521955&vid=331add73efa00881381cbace038e21cbca76318f&prid=web&clid=SAPDAM