α-Terpinene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene
86473-1ML
86473-5ML
SIAL86473-1MLEA
156
EUR
SIAL86473-1ML
SIAL86473-5ML
α-Terpinene, Sigma-Aldrich®
α-Terpinene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 173 °C (1013 hPa) Melting Pt: –48 °C Density: 0.834 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00001534 CAS Number: 99-86-5 EINECS: 202-795-1 UN: 2319 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...