γ-Terpinene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: gamma-Terpinene, p-Mentha-1,4-diene

M-Clarity™ quality level = MQ200

86478-100ML 86478-500ML
SIAL86478-100MLEA 72 EUR
SIAL86478-100ML SIAL86478-500ML
γ-Terpinene, Sigma-Aldrich®
γ-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 182 °C (1013 hPa)
Density: 0.846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001537
CAS Number: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR