γ-Terpinene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: gamma-Terpinene, p-Mentha-1,4-diene
M-Clarity™ quality level = MQ300
W355909-1KG-K W355909-20KG-K W355909-8KG-K
SIALW355909-1KG-KEA 84.6 EUR
SIALW355909-1KG-K SIALW355909-20KG-K SIALW355909-8KG-K
γ-Terpinene, Sigma-Aldrich®
γ-Terpinene
Formula: C₁₀H₁₆
Boiling Pt: 182 °C (1013 hPa)
Density: 0.846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001537
CAS Number: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III