Cyclobutanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
C96001-1G
C96001-5G
SIALC96001-1GEA
60
EUR
SIALC96001-1G
SIALC96001-5G
Cyclobutanone, Sigma-Aldrich®
Cyclobutanone
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 99 Density: 0.93 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001332 CAS Number: 1191-95-3 EINECS: 214-745-6 UN: 1224 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...