Cycloheptanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C99000-100G
C99000-250G
C99000-25G
SIALC99000-100GEA
134
EUR
SIALC99000-100G
SIALC99000-250G
SIALC99000-25G
Cycloheptanone, Sigma-Aldrich®
Cycloheptanone
Formula:
C₇H₁₂O Boiling Pt: 179 °C (1013 hPa) Melting Pt: –21 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004159 CAS Number: 502-42-1 EINECS: 207-937-6 UN: 1224 ADR: 3,III Merck Index: 13,02751 |