Cyclooctanone, Sigma-Aldrich®
Supplier: Merck
Danger
MQ200
C109800-100G C109800-25G
SIALC109800-100GEA 114 EUR
SIALC109800-100G SIALC109800-25G
Cyclooctanone, Sigma-Aldrich®
Cyclooctanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 196
Melting Pt: 39…41 °C
Density: 0.9581 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001754
CAS Number: 502-49-8
EINECS: 207-940-2
UN: 1759
ADR: 8 III