Cyclobutanecarbonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cyclobutanecarboxylic acid chloride, Cyclobutanecarboxylic chloride, Cyclobutanecarbonylchloride
M-Clarity™ quality level = MQ100
C95706-10G C95706-50G
SIALC95706-10GEA 99.8 EUR
SIALC95706-10G SIALC95706-50G
Cyclobutanecarbonyl chloride, Sigma-Aldrich®
Cyclobutanecarbonyl chloride
Formula: C₅H₇ClO
Boiling Pt: 60 °C (1013 hPa)
Storage Temperature: Refrigerator
MDL Number: MFCD00001319
CAS Number: 5006-22-4
EINECS: 225-680-8
UN: 2920
ADR: 8,II