Cyclobutanecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
C95609-100G C95609-25G C95609-5G
SIALC95609-100GEA 173 EUR
SIALC95609-100G SIALC95609-25G SIALC95609-5G
Cyclobutanecarboxylic acid, Sigma-Aldrich®
Cyclobutanecarboxylic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001323
CAS Number: 3721-95-7
EINECS: 223-072-7
UN: 3265
ADR: 8,III