Cyclohexanecarboxylic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Hexahydrobenzoic acid

M-Clarity™ quality level = MQ100

101834-100G 101834-500G 101834-5G
SIAL101834-100GEA 45.7 EUR
SIAL101834-100G SIAL101834-500G SIAL101834-5G
Cyclohexanecarboxylic acid, Sigma-Aldrich®
Cyclohexanecarboxylic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 232
Melting Pt: 31
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5
EINECS: 202-711-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR