Cyclopentanecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
C112003-100G
C112003-25G
C112003-5G
SIALC112003-100GEA
220
EUR
SIALC112003-100G
SIALC112003-25G
SIALC112003-5G
Cyclopentanecarboxylic acid, Sigma-Aldrich®
Cyclopentanecarboxylic acid
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 216 °C (1013 hPa) Melting Pt: 4 °C Density: 1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001371 CAS Number: 3400-45-1 EINECS: 222-269-5 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...