Cyclopropanecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
C116602-100G
C116602-25G
SIALC116602-100GEA
56.7
EUR
SIALC116602-100G
SIALC116602-25G
Cyclopropanecarboxylic acid, Sigma-Aldrich®
Cyclopropanecarboxylic acid
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 17…19 °C Density: 1.087 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001287 CAS Number: 1759-53-1 EINECS: 217-162-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...