2,2,3,3-Tetrafluorocyclobutanol 95%
Supplier: Thermo Scientific
|
Warning
|
L19186.03
L19186.06
L19186.03EA
107
EUR
L19186.03
L19186.06
2,2,3,3-Tetrafluorocyclobutanol 95%
2,2,3,3-Tetrafluorocyclobutanol
Purity: 95%
CAS No.: 374-32-3
Molecular Formula: C4H4F4O
Formula Weight: 144.07
Flash Point: 44°(111°F)
Refractive Index: 1.3680
Boiling Point: 155-157°
Beilstein Reference: 2429286
UN No.: 1987
MDL No.: MFCD00190085
Formula:
C₄H₄F₄O MW: 144.07 g/mol Boiling Pt: 155…157 °C Melting Pt: –22 °C Density: 1.414 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190085 CAS Number: 374-32-3 UN: 1987 ADR: 3,III |