Cyclohexylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aminocyclohexane
M-Clarity™ quality level = MQ200
240648-100ML 240648-5ML
SIAL240648-100MLEA 36.3 EUR
SIAL240648-100ML SIAL240648-5ML
Cyclohexylamine, Sigma-Aldrich®
Cyclohexylamine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 134.5 °C (1013 hPa)
Melting Pt: –18 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
EINECS: 203-629-0
UN: 2357
ADR: 8,II
Merck Index: 13,02758