(Bromomethyl)cyclobutane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Cyclobutyl methyl bromide
M-Clarity™ quality level = MQ100
441171-25ML
441171-5ML
SIAL441171-25MLEA
352
EUR
SIAL441171-25ML
SIAL441171-5ML
(Bromomethyl)cyclobutane, Sigma-Aldrich®
(Bromomethyl)cyclobutane
Formula:
C₅H₉Br Boiling Pt: 123…124 °C (1013 hPa) Density: 1.35 g/cm³ (20 °C) Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040960 CAS Number: 17247-58-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...