3,3,5-Trimethylcyclohexanone
Supplier: Thermo Scientific
BTBG00138.250
BTBG00138.1
MAYBBTBG00138.250EA
0
EUR
MAYBBTBG00138.250
MAYBBTBG00138.1
3,3,5-Trimethylcyclohexanone
3,3,5-Trimethylcyclohexanone
Formula:
C₉H₁₆O MW: 140.23 g/mol Boiling Pt: 188,8 °C (1013 hPa) Melting Pt: –10 °C Density: 0,89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00019466 CAS Number: 873-94-9 EINECS: 212-855-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...